missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMPRSS2 Monoclonal Antibody (A6D9)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA544980
Description
TMPRSS2 Monoclonal Antibody for Western Blot, ICC/IF
TMPRSS2 is a protein that belongs to the serine protease family. The protein contains a type II transmembrane domain, a receptor class A domain, a scavenger receptor cysteine-rich domain and a protease domain. Serine proteases are known to be involved in many physiological and pathological processes. Its gene was demonstrated to be up-regulated by androgenic hormones in prostate cancer cells and down-regulated in androgen-independent prostate cancer tissue. The protease domain of this protein is thought to be cleaved and secreted into cell media after autocleavage.Identifiants chimiques
| TMPRSS2 | |
| 2 mg/mL | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O15393 | |
| 7113 | |
| Synthetic peptide within human TMPRSS2 aa 1-50. | |
| Antibody |
| A6D9 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG1 | |
| Human | |
| TMPRSS2 | |
| D16Ertd61e; EC 3.4.219; epitheliasin; FLJ41954; MGC6821; Plasmic transmembrane protein X; PP9284; PRSS10; Serine protease 10; Tmprss2; TMPS2; Transmembrane protease serine 2; Transmembrane protease serine 2 catalytic chain; Transmembrane protease serine 2 non-catalytic chain; transmembrane protease, serine 2 | |
| TMPRSS2 | |
| Primary | |
| Protein G |
Spécifications
| TMPRSS2 | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O15393 | |
| TMPRSS2 | |
| Synthetic peptide within human TMPRSS2 aa 1-50. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot, Immunocytochemistry | |
| A6D9 | |
| Unconjugated | |
| TMPRSS2 | |
| D16Ertd61e; EC 3.4.219; epitheliasin; FLJ41954; MGC6821; Plasmic transmembrane protein X; PP9284; PRSS10; Serine protease 10; Tmprss2; TMPS2; Transmembrane protease serine 2; Transmembrane protease serine 2 catalytic chain; Transmembrane protease serine 2 non-catalytic chain; transmembrane protease, serine 2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 7113 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |