missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TM2 TEAD inhibitor
Description
TM2 TEAD inhibitor is a potent and reversible TEA domain transcription factor inhibitor (IC50 = 38 nM and 156 nM for TEAD4 auto-palmitoylation and TEAD2 palmitoylation, respectively). It inhibits TEAD-YAP association and transcriptional activity. TM2 TEAD inhibitor exhibits strong antiproliferation effects as a single agent or in combination with a MEK inhibitor in YAP-dependent cancer cells. Also inhibits YAP-dependent liver organoid growth ex vivo.
Specifications
Specifications
| Chemical Name or Material | 4-[3-(2-Cyclohexylethoxy)benzoyl]-N-phenyl-1-piperazinecarboxamide |
| CAS | 1008768-41-9 |
| Quantity | 50 mg |
| Target | YAP1 |
| Molecular Formula | C26H33N3O3 |
| Purity | 0.98 |