missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titration Solvent, for Neutralization Number of Petroleum Products, Ricca Chemical
Note: May not be shipped via UPS Air Services
$523.20 - $523.20
Specifications
| Chemical Name or Material | Titration Solvent |
|---|---|
| CAS | 108-88-3 |
| Packaging | Amber Glass Bottle |