missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titration Solvent, for Neutralization Number of Petroleum Products, Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Titration Solvent |
| CAS | 108-88-3 |
| Molecular Formula | C7H8 |
| MDL Number | MFCD00008512 |
| Synonym | methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene |
| InChI Key | YXFVVABEGXRONW-UHFFFAOYSA-N |
| SMILES | CC1=CC=CC=C1 |
| IUPAC Name | toluene |
| Molecular Weight (g/mol) | 92.14 |
| Grade | ASTM |
| Show More |