missing translation for 'onlineSavingsMsg'
Learn More

Titration Solvent Blend D2896, Solstice

Catalog No. 60090598
Change view
Click to view available options
19 l
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
60090598 19 l
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60090598 Supplier Honeywell Inc Supplier No. WP215119P

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

LabReady™ Solvent Blend, Total Base Number Titration Solvent (ASTM D 2896)

Specifications

CAS 108-90-7, 64-19-7
CAS Max % 67.90%, 32.10%
Quantity 19 L
Flash Point 7.2°C
Packaging Winpak
Physical Form Liquid
Purity Grade Notes Custom Labready™ blends
Boiling Point 115.6°C