missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titrant 5, AQUASTAR™, EMD Chemicals
Specifications
Specifications
| Color | Red to Brown |
| Density | 0.850-0.860g/cm3 |
| Physical Form | Liquid |
| Quantity | 1 L |
| Packaging | GL45 Poly coated glass bottle |