missing translation for 'onlineSavingsMsg'
Learn More

Titanium Standard For ICP, 1000μg/mL in H2O/0.24 F, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC PLTI92T

 View more versions of this product

Catalog No. PLTI92T


Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ti1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Titanium Standard
Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
0.1,20.0,79.9
0.1,20.0,79.9
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
H2O/0.24 F
Assurance
Triple-leached LDPE bottle
16962-40-6 , 7732-18-5 , 7732-18-5
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
AA, ICP
Certified Reference Material
250 mL
Matrix    Concentration    Cat. No.
H2O/tr 2.4%F-     1000μg/mL    PLTI9-2Y, -2T, -2X
40% HCl    1000μg/mL    PLTI5-2X
40% HCl    10,000μg/mL    PLTI5-3X
H2O/tr 2.4%F-    10,000μg/mL    PLTI9-3Y, -3X

Chemical Identifiers

16962-40-6
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Causes skin irritation, serious eye irritation

DOTInformation : Nonhazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only