missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium Standard For ICP, 10,000μg/mL in H2O/0.24 F, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$428.78 - $1045.91
Chemical Identifiers
| CAS | 16962-40-6 |
|---|
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Ti | 10,000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
| 16962-40-6 |
Specifications
| Triple-leached LDPE bottle | |
| 16962-40-6 , 7732-18-5 | |
| AA, ICP | |
| Certified Reference Material | |
| 125 mL | |
| Titanium Standard |
| Assurance Certificate of Analysis reports up to 30 elements at ppb levels. | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| H2O/2.4% F | |
| Assurance |
Safety and Handling
Causes skin irritation, serious eye irritation
DOTInformation : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only