missing translation for 'onlineSavingsMsg'
Learn More

Titanium sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.995% (metals basis)

Catalog No. AA43042KS
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43042KS 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA43042KS Supplier Thermo Scientific Chemicals Supplier No. 043042KS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-32-6 | Ti | 47.87 g/mol

Titanium sputtering target and its alloys are characterized by their lightness, strength and corrosion resistance and are used widely in aerospace applications. In addition, these properties also make the material suitable for medical applications (e.g. replacement hip joints). Titanium dioxide is used as a white pigment in paints and plastics as it provides great opacity. The same material is also used in the manufacture of heat resisting and durable glass, the TiO2 replacing certain proportions of the soda. Titanium carbide is used to manufacture cemented carbides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-32-6
Molecular Formula Ti
Molecular Weight (g/mol) 47.87
MDL Number MFCD00011264,MFCD00151301
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
Synonym oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]

Specifications

Quantity 1 Ea.
Odor Odorless
Solubility Information Insoluble in water.
Percent Purity 99.995%
Assay (metals basis)
Chemical Name or Material Titanium sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
EINECSNumber 231-142-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only