missing translation for 'onlineSavingsMsg'
Learn More

Titanium slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, Puratronic™, 99.995% (metals basis)

Catalog No. AA4239414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4239414 25 g
AA4239406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4239414 Supplier Thermo Scientific Chemicals Supplier No. 04239414

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-32-6 | Ti | 47.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-32-6
Molecular Formula Ti
Molecular Weight (g/mol) 47.87
MDL Number MFCD00011264,MFCD00151301
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
Synonym oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]

Specifications

Melting Point 1660°C
Density 4.5 g/mL
Quantity 25 g
Odor Odorless
Weight ≈0.11g/slug
Form Slug
Percent Purity 99.995%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Titanium slug, 3.175mm (0.125 in.) dia. x 3.175mm (0.125 in.) length
EINECSNumber 231-142-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only