missing translation for 'onlineSavingsMsg'
Learn More

Titanium Dioxide, Powder, Reagents

Catalog No. 01336929
Change view
Click to view available options
Quantity:
500 g
12 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336929 12 kg
01336928 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01336929 Supplier Labchem Inc Supplier No. C234050012F4

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Titanium Dioxide, Powder, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 13463-67-7
Molecular Formula O2Ti
Molecular Weight (g/mol) 79.87
MDL Number MFCD00011269 MFCD00210650
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
IUPAC Name dioxotitanium
SMILES O=[Ti]=O

Specifications

Boiling Point 2500°C
Melting Point 1800°C
Color White
Physical Form Solid
Quantity 12 kg
Percent Purity 100%
Grade Reagent
Packaging Fiber Drum
Specific Gravity 4.2
Chemical Name or Material Titanium Dioxide, Powder