Learn More
Fisherbrand™ Tissue Path™ II Cassettes
Standard lids fit securely onto cassettes.
Supplier:  Fisherbrand™ 58129
 Afficher plus de versions de ce produit
Numéro de catalogue. 15-120-400C
Il en reste null
Description
Description
  • Molded from a special high-density polymer resistant to all commonly used histological solvents
  • Slanted writing surface accepts markings easily, permitting positive sample identification
Order Info
Packaged in three dispenser boxes of 500
Spécifications
Spécifications
Separate Lid
Green
High Density Polymer
Tissue Cassette
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS