missing translation for 'onlineSavingsMsg'
Learn More

TISAB II, For Fluoride, Certified, LabChem™

Catalog No. LC261302
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC261302 1 L
LC261304 4 L
LC261306 10 L
LC261305 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. LC261302 Supplier LabChem Supplier No. LC261302

Specifications

Chemical Name or Material TISAB II
Color Colorless
Name Note For Fluoride
CAS Max % 83.7, 5.8, 5.7, 4.4, 0.4
CAS 7647-14-5, 64-19-7, 1310-73-2, 125572-95-4
Linear Formula CH3COOH
Solubility Information Soluble in water
Formula Weight 60.05
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Traceability to NIST Traceable to NIST
Decomposition Information Carbon monoxide; Carbon dioxide
pH 5.3 to 5.5
Quantity 1 L
Physical Form Liquid
Show More Show Less