missing translation for 'onlineSavingsMsg'
Learn More

TISAB I, for Fluoride ISE, Reagents

Catalog No. 01336225
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336225 1 L
01336224 20 L
01336223 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01336225 Supplier Labchem Inc Supplier No. C73402001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

TISAB I, for Fluoride ISE, Reagent, use for Fluoride analysis. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 64-19-7 , 64-19-7
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Total Ionic Strength Adjustment Buffer
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless
For Use With (Application) Fluoride Analysis
For Use With (Equipment) Fluoride Meter
pH 5 to 5.5
Physical Form Liquid
Quantity 1 L
Solution Type Buffer
Packaging Poly Bottle
Specific Gravity 1.1
Sterility Non-sterile
Grade Reagent
Chemical Name or Material TISAB I, for Fluoride ISE
Show More Show Less