missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TISAB with CDTA (TISAB II), for Fluoride ISE, Reagents
Specifications
Specifications
| Boiling Point | 100°C |
| Color | Colorless |
| For Use With (Application) | Fluoride Analysis |
| For Use With (Equipment) | Fluoride Meter |
| pH | 5 to 5.5 |
| Physical Form | Liquid |
| Quantity | 10 L |
| Solution Type | Buffer |
| Packaging | Cubitainer |
| Specific Gravity | 1.0343 |
| Show More |