missing translation for 'onlineSavingsMsg'
Learn More

Tinidazole 98.0+%, TCI America™

Catalog No. T30585G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30585G 5 g
T305825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T30585G Supplier TCI America Supplier No. T30585G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 19387-91-8 | C8H13N3O4S | 247.27 g/mol

Chemical Identifiers

CAS 19387-91-8
Molecular Formula C8H13N3O4S
Molecular Weight (g/mol) 247.27
MDL Number MFCD00057217
InChI Key HJLSLZFTEKNLFI-UHFFFAOYSA-N
Synonym 1-[2-(Ethylsulfonyl)ethyl]-2-methyl-5-nitroimidazole
PubChem CID 5479
ChEBI CHEBI:63627
IUPAC Name 1-[2-(ethanesulfonyl)ethyl]-2-methyl-5-nitro-1H-imidazole
SMILES CCS(=O)(=O)CCN1C(C)=NC=C1[N+]([O-])=O

Specifications

Melting Point 129°C
Color White-Yellow
Quantity 5 g
Formula Weight 247.27
Percent Purity ≥98.0% (HPLC,T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material Tinidazole
RTECSNumber NI6255000
TSCA No