missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) Phthalocyanine, TCI America™

Catalog No. P10241G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P10241G Supplier TCI America Supplier No. P10241G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15304-57-1
Molecular Formula C32H16N8Sn
Molecular Weight (g/mol) 631.25
MDL Number MFCD00058867
InChI Key LLVONELOQJAYBZ-UHFFFAOYSA-N
Synonym tin ii phthalocyanine, tin phthalocyanine, snpc, phthalocyanatotin cento

Specifications

Color Black-Purple
Physical Form Crystalline Powder
Quantity 1 g
UN Number 3146
Formula Weight 631.25
Chemical Name or Material Tin(II) Phthalocyanine
EINECSNumber (5)-4129
TSCA No