missing translation for 'onlineSavingsMsg'
Learn More

Tin ICP-MS Standard, 1000 ppm Sn in 3% HNO3, Ricca Chemical

Catalog No. MSSN1KN100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MSSN1KN100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MSSN1KN100 Supplier Ricca Chemical Company Supplier No. MSSN1KN100

Chemical Identifiers

CAS 7440-31-5 , 7697-37-2 , 7664-39-3 , 7732-18-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Sn
CAS Min % 93.0, 4.89, 0.12, 0.09
CAS Max % 97.0, 5.09, 0.12, 0.1
Equivalence 1 mL = 1 mg Sn
Physical Form Liquid
Color Colorless
pH < 2
Melting Point Approximately 0°C
Boiling Point Approximately 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type ICP Single-element Standard Solution
Quantity 100 mL
Grade Laboratory
Chemical Name or Material Tin ICP-MS Standard
Show More Show Less