missing translation for 'onlineSavingsMsg'
Learn More

Tin AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg Sn), LabChem™

Catalog No. lc261001
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC261001 500 mL Poly Bottle
LC261007 125 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC261001 Supplier LabChem Supplier No. LC261001
Only null left

Chemical Identifiers

CAS 10025-69-1 , 7647-01-0 , 7732-18-5

Specifications

Packaging Poly Bottle
CAS Max % 79.83, 19.98, 0.19
Identification Passes Test
Viscosity 1.25cSt
Density 1.1g/mL
Physical Form Liquid
Concentration 1000 ppm ±10 ppm (1mL = 1mg Sn)
Linear Formula SnCl2·2H2O
UN Number UN1789
Color Colorless
Solubility Soluble in water
Quantity 500 mL
Formula Weight 225.65
Grade Certified
Chemical Name or Material Tin AA Standard, 1000ppm (1mL = 1mg Sn)
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Store in a well-ventilated place.
Keep container tightly closed.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature