missing translation for 'onlineSavingsMsg'
Learn More

Tin AA Standard, certifié, 1000 ppm ±10 ppm (1 mL = 1 mg Sn), LabChem™

Numéro de catalogue. LC261007
Modifier l'affichage
Click to view available options
Quantité:
125 mL
500 mL
Emballage:
Bouteille en polyéthylène
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
LC261007 125 mL Bouteille en polyéthylène
LC261001 500 mL Bouteille en polyéthylène
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. LC261007 Fournisseur LabChem Code fournisseur LC261007

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10025-69-1 , 7647-01-0 , 7732-18-5

Specifications

Nom ou substance chimique Tin AA Standard
Emballage Bouteille en polyéthylène
Nom Remarque 1000ppm (1mL = 1mg Sn)
CAS % maximal 79.83, 19.98, 0.19
Désignation Test de réussite
Viscosité 1.25cSt
Densité 1.1g/mL
Forme physique Liquide
Concentration 1000 ppm ±10 ppm (1 ml = 1 mg Sn)
Formule linéaire SnCl2·2H2O
Numéro UN UN1789
Couleur Incolore
Solubilité Soluble dans l’eau
Quantité 125 mL
Poids de formule 225.65
Niveau Certifié
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Store in a well-ventilated place.
Keep container tightly closed.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Température ambiante