missing translation for 'onlineSavingsMsg'
Learn More

PIG™ Tight-Head UN Rated 5 Gal. Carbon Steel Drum

Catalog No. 18366512
Change view
Click to view available options
Product Type:
Lined Drum
Unlined Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
18366512 Unlined Drum
18366511 Lined Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18366512 Supplier PIG™ Supplier No. DRM1427BKWULD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tough, UN Rated drums that are made for hazardous materials.

  • UN Rated and tested to help ensure safe shipping of hazardous materials
  • Tight-head design is equipped with 2 in. NPT and 3/4 in. NPT bungs
  • Choose optional phenolic/epoxy lining for added corrosion resistance
  • Steel lid, body and bottom meet strict wall thickness requirements to ensure strength, durability and long life

Specifications

Product Type Unlined Drum
Capacity (English) 55 gal.
Capacity (Metric) 208.2 L
Color Black
Material Carbon Steel
Height (English) 35.25 in.
Height (Metric) 90 cm
Diameter (English) Inner 22.5 in.
Diameter (English) Opening 57.15 cm
Diameter (English) Outer 23 in.
Diameter (Metric) Inner 58 in.
Show More Show Less