missing translation for 'onlineSavingsMsg'
Learn More

Ticlopidine hydrochloride, ≥99%, MilliporeSigma™ Supelco™

$78.87 - $78.87

Specifications

CAS 53885-35-1
Physical Form Neat
Linear Formula C14H14 ClNS · HCl
MDL Number MFCD00079606
Synonym 5-(o-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111014526
View Documents
Merck Emd Millipore
T66541G
1 g
Each for $78.87
Only null left
 
Description

Description

Featured Industries: Pharmaceutical (small molecule)
Specifications

Specifications

53885-35-1
C14H14 ClNS · HCl
5-(o-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine
Drug Analysis
≥99%
Neat
MFCD00079606
Analytical Standard
300.25
Analytical Standard
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'