missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tiaprofenic acid

$268.36 - $817.65

Chemical Identifiers

CAS 33005-95-7
Molecular Formula C14H12O3S
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC467170010
View Documents
Thermo Scientific Chemicals
467170010
1 g
Each for $817.65
Only null left
 
AC467172500
View Documents
Thermo Scientific Chemicals
467172500
250 mg
Each for $268.36
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

33005-95-7
C14H12O3S

Specifications

33005-95-7
95.0°C to 100.0°C
260.31
CC(C(O)=O)C1=CC=C(S1)C(=O)C1=CC=CC=C1
260.31
1 g
C14H12O3S
GUHPRPJDBZHYCJ-UHFFFAOYNA-N
2-(5-benzoylthiophen-2-yl)propanoic acid
Tiaprofenic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement Toxic if swallowed.

Causes damage to organs through prolonged or repeated exposure if inhaled.

GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.

IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Store locked up.

Do not breathe dust/fume/gas/mist/vapors/spray.

Get medical attention/advice if you feel unwell.

GHS Signal Word: Danger

EINECSNumber : 251-329-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only