missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tiaprofenic acid

Catalog No. AC467170010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467170010 1 g
AC467172500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467170010 Supplier Thermo Scientific Chemicals Supplier No. 467170010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33005-95-7
Molecular Formula C14H12O3S

Specifications

Quantity 1 g
Melting Point 95.0°C to 100.0°C
Formula Weight 260.31
InChI Key GUHPRPJDBZHYCJ-UHFFFAOYNA-N
SMILES CC(C(O)=O)C1=CC=C(S1)C(=O)C1=CC=CC=C1
IUPAC Name 2-(5-benzoylthiophen-2-yl)propanoic acid
Molecular Weight (g/mol) 260.31
Chemical Name or Material Tiaprofenic acid
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Toxic if swallowed.

Causes damage to organs through prolonged or repeated exposure if inhaled.

Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.

IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Store locked up.

Do not breathe dust/fume/gas/mist/vapors/spray.

Get medical attention/advice if you feel unwell.

EINECSNumber 251-329-3

RUO – Research Use Only