missing translation for 'onlineSavingsMsg'
Learn More

Tians epic™ Basic Protection Cleanroom Frocks

Low-lint frock features knit wrists and collar for use in controlled environments.

$182.00 - $189.40

Specifications

Product Type Controlled Environments Frock
Material Spunbond Polypropylene
Color White
Disposable Reusable
Sterility Non-sterile
View More Specs

Products 4
Catalog Number Mfr. No. Size Length (English) Length (Metric) Price Quantity  
Catalog Number Mfr. No. Size Length (English) Length (Metric) Price Quantity  
19-038-761
View Documents
Tians
843885NP2XL
2X-Large 20.4 in. 51.8 cm N/A
Only null left
 
19-038-762
View Documents
Tians
843885NP3XL
3X-Large 16.5 in. 41.9 cm N/A
Only null left
 
19-038-760
View Documents
Tians
843885NPXL
X-Large 20.4 in. 51.8 cm N/A
Only null left
 
19-038-764
View Documents
Tians
843885NP5X
5X-Large N/A
Only null left
 
Description

Description

  • Latex free, low-lint frock for use in controlled environments and many general application areas
  • Breathable cloth-like fabric is comfortable and cool
  • Plastic snaps
  • Knit wrists and knit collar
  • No pockets
  • Constructed from Environstar-SPP spunbonded polypropylene fabric
  • Neatly folded and cleanroom packaged
  • 10 pieces per inner bag, 5 inner bags per master bag, 50 pieces per case
Specifications

Specifications

Controlled Environments Frock
White
Non-sterile
Knit
Yes
Spunbond Polypropylene
Reusable
Snaps
Knit
Class 10,000 (ISO 7)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'