missing translation for 'onlineSavingsMsg'
Learn More

Thymolphthalein, TCI America™

Catalog No. T02371G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T02371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T02371G Supplier TCI America Supplier No. T02371G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 125-20-2
Molecular Formula C28H30O4
Molecular Weight (g/mol) 430.544
MDL Number MFCD00005909
InChI Key LDKDGDIWEUUXSH-UHFFFAOYSA-N
PubChem CID 31316
IUPAC Name 3,3-bis(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-2-benzofuran-1-one
SMILES CC1=CC(=C(C=C1C2(C3=CC=CC=C3C(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O

Specifications

Melting Point 253°C
Color White/Yellow
Quantity 1 g
Physical Form Crystal/Powder
Formula Weight 430.54
Chemical Name or Material Thymolphthalein