missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thymolphthalein Indicator, 0.1% (w/v) in 90% (v/v) Isopropyl Alcohol, Ricca Chemical
Specifications
Specifications
| Odor | Characteristic alcohol odor |
| Packaging | Natural Poly Bottle |
| Solvent or Matrix | Aqueous Solution |
| Color | Colorless |
| Solubility Information | Solubility in Water: Infinite |
| Quantity | 1 L |
| Percent Purity | 0.1% W/V |
| Grade | Indicator |
| Chemical Name or Material | Thymolphthalein Indicator, 0.1% (w/v) in 90% (v/v) Isopropyl Alcohols |