missing translation for 'onlineSavingsMsg'
Learn More

Thymolphthalein, ACS Reagent Grade, Ricca Chemical

Catalog No. RDCT045025F
Change view
Click to view available options
Package: Pouch; Size: 25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
RDCT045025F Package: Pouch; Size: 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCT045025F Supplier Ricca Chemical Company Supplier No. RDCT045025F1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 125-20-2
CAS Min % 98.0
CAS Max % 100.0
Molecular Formula C28H30O4
InChI Key LDKDGDIWEUUXSH-UHFFFAOYSA-N
SMILES CC1=CC(=C(C=C1C2(C3=CC=CC=C3C(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O
IUPAC Name 3,3-bis(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-2-benzofuran-1-one
Molecular Weight (g/mol) 430.544
PubChem CID 31316
Grade ACS Reagent
Packaging Pouch
Quantity 25 g
Chemical Name or Material Thymolphthalein
Show More Show Less
ShelfLife 60 months