missing translation for 'onlineSavingsMsg'
Learn More

Thymolphthalein, 0.1% (w/v) in Methanol, Ricca Chemical

Catalog No. 845532
Change view
Click to view available options
Quantity:
1 L
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
845532 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 845532 Supplier Ricca Chemical Company Supplier No. 845532

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 125-20-2 , 67-56-1
Molecular Formula C28H30O4
Molecular Weight (g/mol) Mixture
InChI Key LDKDGDIWEUUXSH-UHFFFAOYSA-N
PubChem CID 31316
IUPAC Name 3,3-bis(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-2-benzofuran-1-one
SMILES CC1=CC(=C(C=C1C2(C3=CC=CC=C3C(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O

Specifications

CAS Min % 98.0, 0.12
CAS Max % 100.0, 0.13
Concentration 0.1% (w/v)
Packaging Natural Poly Bottle
Solvent or Matrix Methanol
Color Colorless
Quantity 1 L
Physical Form Liquid
Percent Purity 0.1% W/V
Grade Indicator
Chemical Name or Material Thymolphthalein
Show More Show Less