missing translation for 'onlineSavingsMsg'
Learn More

Thulium AA Standard, 1000 ppm Tm in 3% HNO3, Ricca Chemical

Catalog No. p-3340526
Change view
Click to view available options
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
ATM1KN100 Natural LDPE Bottle
ATM1KN500 Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ATM1KN100 Supplier Ricca Chemical Company Supplier No. ATM1KN100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 12036-44-1 , 7697-37-2 , 7732-18-5

Specifications

Packaging Natural LDPE Bottle
CAS Min % 93.0, 4.56, 0.11
CAS Max % 97.0, 4.74, 0.11
Odor Odorless
Concentration or Composition (by Analyte or Components) 1mL = 1mg Tm (1, 000ppm Tm)Tm2O3 in 3% HNO3
Chemical Name or Material Thullium AA Standard
ShelfLife 24 months