missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thulium AA Standard, 1000 ppm Tm in 3% HNO3, Ricca Chemical
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| CAS Min % | 93.0, 4.56, 0.11 |
| CAS Max % | 97.0, 4.74, 0.11 |
| Odor | Odorless |
| Concentration or Composition (by Analyte or Components) | 1mL = 1mg Tm (1, 000ppm Tm)Tm2O3 in 3% HNO3 |
| Chemical Name or Material | Thullium AA Standard |
Safety and Handling
| ShelfLife | 24 months |