missing translation for 'onlineSavingsMsg'
Learn More

Thulium(III) Trifluoromethanesulfonate 88.0+%, TCI America™

Catalog No. T19205G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T19205G Supplier TCI America Supplier No. T19205G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 141478-68-4
Molecular Formula C3F9O9S3Tm
Molecular Weight (g/mol) 616.124
MDL Number MFCD00209617
InChI Key PBASUZORNBYVFM-UHFFFAOYSA-K
Synonym thulium iii trifluoromethanesulfonate, thulium iii triflate, trifluoromethanesulfonic acid thulium iii salt, thulium triflate, tm otf 3, acmc-1c8m6, thulium 3+ tritriflate, thulium 3+ ; trifluoromethanesulfonate, thulium trifluoromethanesulfonate, thulium iii tris trifluoromethanesulfonate
PubChem CID 2733936
IUPAC Name thulium(3+);trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Tm+3]

Specifications

Color White
Quantity 5 g
UN Number 1759
Formula Weight 616.12
Percent Purity ≥88.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Thulium(III) Trifluoromethanesulfonate