missing translation for 'onlineSavingsMsg'
Learn More

Thiourea, ACS, 99% min.

Catalog No. AA36609A1
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA36609A1 1 kg
AA3660918 50 g
AA3660930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA36609A1 Supplier Thermo Scientific Chemicals Supplier No. 036609A1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reagent for organic synthesis | CAS: 62-56-6 | CH4N2S | 76.12 g/mol

Used as a reagent for organic synthesis. Thiourea is a photographic fixative, and used in manufacture of resins. It act as a catalyst for asymmetric reactions. It plays an essential role as catalyst for highly enantio- and diastereoselective additions reaction of oxindoles to nitroolefins. It is also useful to improve the productivity of mung bean.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62-56-6
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.12
MDL Number MFCD00008067
InChI Key UMGDCJDMYOKAJW-UHFFFAOYSA-N
Synonym thiocarbamide, 2-thiourea, pseudothiourea, isothiourea, sulourea, 2-thiopseudourea, sulfourea, thiuronium, thiocarbonic acid diamide, urea, thio
PubChem CID 2723790
ChEBI CHEBI:36946
IUPAC Name thiourea
SMILES NC(N)=S

Specifications

Melting Point 175°C to 178°C
Color White
Density 1.405 g/mL
Odor Odorless
Linear Formula H2NCSNH2
Quantity 1 kg
UN Number UN3077
Beilstein 605327
Merck Index 14,9367
Solubility Information Soluble in water.
Formula Weight 76.12
Percent Purity ≤99%
Grade ACS
Physical Form Crystalline
Chemical Name or Material Thiourea
Show More Show Less
Hazard Category H302-H350-H361d-H500
Hazard Statement GHS H Statement
H351-H361-H302
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 200-543-5
RTECSNumber YU2800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only