missing translation for 'onlineSavingsMsg'
Learn More

Thiosemicarbazide, 98%

Catalog No. AAA1463022
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1463022 100 g
AAA1463036 500 g
AAA146300E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1463022 Supplier Thermo Scientific Chemicals Supplier No. A1463022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 79-19-6 | CH5N3S | 91.132 g/mol

Used in TLC to stain alpha-keto acidsThiosemicarbazide is used as a reagent for ketones as well as chemical intermediate. It is involved in the detection of metals. It is also used in photography. Further, it is effective for controlling bacterial leaf blight in rice. In addition to this, it is used to prepare 1,3,4-thiadiazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in TLC to stain alpha-keto acidsThiosemicarbazide is used as a reagent for ketones as well as chemical intermediate. It is involved in the detection of metals. It is also used in photography. Further, it is effective for controlling bacterial leaf blight in rice. In addition to this, it is used to prepare 1,3,4-thiadiazoles.

Solubility
Soluble in water, ethanol, alcohols and hot methanol.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.

Chemical Identifiers

CAS 79-19-6
Molecular Formula CH5N3S
Molecular Weight (g/mol) 91.132
MDL Number MFCD00007620
InChI Key BRWIZMBXBAOCCF-UHFFFAOYSA-N
Synonym thiosemicarbazide, hydrazinecarbothioamide, n-aminothiourea, 1-aminothiourea, semicarbazide, thio, isothiosemicarbazide, 2-thiosemicarbazide, 3-thiosemicarbazide, thiocarbamylhydrazine, thiocarbamoylhydrazine
PubChem CID 2723789
IUPAC Name aminothiourea
SMILES C(=S)(N)NN

Specifications

Melting Point ∼180°C (decomposition)
Odor Odorless
Quantity 100 g
UN Number UN2811
Beilstein 506320
Merck Index 14,9361
Solubility Information Soluble in water,ethanol,alcohols and hot methanol.
Formula Weight 91.14
Percent Purity 99%
Chemical Name or Material Thiosemicarbazide
Hazard Category H300
Hazard Statement GHS H Statement
H300
Fatal if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 201-184-7
RTECSNumber VT4200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only