missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiophosgene, 85%, AcroSeal™, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécifications
Spécifications
| Nom ou substance chimique | Thiophosgene |
| Quantité | 100mL |
| Fourchette de pourcentages d’essai | 80% min. (GC) |
| Formule linéaire | ClCSCl |
| Point d’éclair | 62°C |
| Renseignements sur la solubilité | Solubility in water: slow decomposition in cold, fast in hot water. Other solubilities: soluble in ether, ccl4, decomposes in alcohol |
| Poids de formule | 114.98 |
| Pourcentage de pureté | 85% |
| Emballage | AcroSeal™ Glass bottle |
| Indice de réfraction | 1.5442 |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. Toxic if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. If skin i |
| EINECSNumber | 207-341-6 |
RUO – Research Use Only