missing translation for 'onlineSavingsMsg'
Learn More

Thiophenol, MilliporeSigma™ Supelco™

Catalog No. 111019943
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019943 5 mL
111018402 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019943 Supplier Merck Emd Millipore Supplier No. 092865MLF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

CAS 108-98-5
Vapor Pressure 1.4 mmHg (20°C)
Vapor Density 3.8 (vs. air)
Density 1.073 g/mL (at 25°C)
Flash Point 50°C
Linear Formula C6H5 SH
MDL Number MFCD00004826
Melting Point -15°C
Boiling Point 169°C
Synonym Benzenethiol; Phenyl mercaptan
Type Analytical Standard
Quantity 5 mL
Formula Weight 110.18
Grade Analytical Standard
Show More Show Less