missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-3-acetonitrile 98.0+%, TCI America™

Catalog No. T11935G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T11935G Supplier TCI America Supplier No. T11935G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13781-53-8
Molecular Formula C6H5NS
Molecular Weight (g/mol) 123.17
MDL Number MFCD00005471
InChI Key GWZCLMWEJWPFFA-UHFFFAOYSA-N
Synonym 3-thiopheneacetonitrile, thiophene-3-acetonitrile, 2-thiophen-3-yl acetonitrile, 3-thienylacetonitrile, 3-cyanomethylthiophene, 3-thiophene acetonitrile, thien-3-ylacetonitrile, 2-3-thienyl acetonitrile, thiophen-3-acetonitrile, 2-3-thienyl ethanenitrile
PubChem CID 83730
IUPAC Name 2-(thiophen-3-yl)acetonitrile
SMILES N#CCC1=CSC=C1

Specifications

Color Yellow
Boiling Point 125°C
Quantity 5 g
UN Number 3276
Formula Weight 123.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Thiophene-3-acetonitrile