missing translation for 'onlineSavingsMsg'
Learn More

Thionyl bromide, 97%

Catalog No. AAL1564814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1564814 25 g
AAL1564822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1564814 Supplier Thermo Scientific Chemicals Supplier No. L1564814

CAS: 507-16-4 | Br2OS | 207.87 g/mol

Thionyl bromide is used as a brominating agent in organic synthesis. It is used in the bromination of alfa-beta-unsaturated carbonyl compounds. It is also used to prepare alkyl bromides from alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thionyl bromide is used as a brominating agent in organic synthesis. It is used in the bromination of alfa-beta-unsaturated carbonyl compounds. It is also used to prepare alkyl bromides from alcohols.

Solubility
Miscible with benzene, toluene and ether.

Notes
Moisture and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with water, hydrobromide, acetone and alcohol.It hydrolyzes readily to release hydrogen bromide.

Chemical Identifiers

CAS 507-16-4
Molecular Formula Br2OS
Molecular Weight (g/mol) 207.87
MDL Number MFCD00011448
InChI Key HFRXJVQOXRXOPP-UHFFFAOYSA-N
Synonym thionyl bromide, thionyl dibromide, unii-n6cu78b13t, thionylbromide, sulfur oxy dibromide, acmc-20ajin, pubchem2023, sobr2, thionyl bromide, purum at
PubChem CID 68176
IUPAC Name sulfurooyl dibromide
SMILES BrS(Br)=O

Specifications

Boiling Point 47°C to 49°C (20mmHg)
Melting Point -52°C
Quantity 25 g
UN Number UN3264
Merck Index 14,9347
Solubility Information Miscible with benzene,toluene and ether.
Formula Weight 207.88
Percent Purity 97%
Odor Acrid
Refractive Index 1.675
Sensitivity Light and moisture sensitive
Density 2.683
Chemical Name or Material Thionyl bromide
Show More Show Less
Hazard Category H312-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: I
EINECSNumber 208-064-3
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only