missing translation for 'onlineSavingsMsg'
Learn More

Thionicotinamide 98.0+%, TCI America™

Catalog No. T076525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T076525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T076525G Supplier TCI America Supplier No. T076525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4621-66-3
Molecular Formula C6H6N2S
Molecular Weight (g/mol) 138.19
MDL Number MFCD00006399
InChI Key XQWBMZWDJAZPPX-UHFFFAOYSA-N
Synonym thionicotinamide, 3-pyridinecarbothioamide, nicotinothioamide, 3-thiocarbamoylpyridine, 3-thioamidopyridine, nicotinamide, thio, 3-pyridylthioformamide, 3-pyridinethiocarboxamide, thio-3-pyridinecarboxamide, unii-6a0hxx0753
PubChem CID 737155
IUPAC Name pyridine-3-carbothioamide
SMILES NC(=S)C1=CC=CN=C1

Specifications

Melting Point 185°C
Quantity 25 g
Formula Weight 138.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Thionicotinamide
RTECSNumber QS4488000
TSCA Yes