missing translation for 'onlineSavingsMsg'
Learn More

Thioglycolic Acid 98.0+%, TCI America™

Catalog No. M0052500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0052500G 500 g
M005225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0052500G Supplier TCI America Supplier No. M0052500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 68-11-1
Molecular Formula C2H4O2S
Molecular Weight (g/mol) 92.112
MDL Number MFCD00004876
InChI Key CWERGRDVMFNCDR-UHFFFAOYSA-N
Synonym mercaptoacetic acid, thioglycolic acid, 2-thioglycolic acid, acetic acid, mercapto, 2-mercaptoacetic acid, sulfanylacetic acid, thiovanic acid, mercaptoessigsaeure, thioglycollic acid, glycolic acid, thio
PubChem CID 1133
ChEBI CHEBI:30065
IUPAC Name 2-sulfanylacetic acid
SMILES C(C(=O)O)S

Specifications

Melting Point -16°C
Color Colorless
Boiling Point 96°C
Quantity 500 g
UN Number 1940
Formula Weight 92.11
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Thioglycolic Acid
EINECSNumber (2)-1355
RTECSNumber AI5950000
TSCA Yes
Recommended Storage Refrigerator