missing translation for 'onlineSavingsMsg'
Learn More

Thioflavine T, tech. 75%

Catalog No. AAJ6104314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6104314 25 g
AAJ6104306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6104314 Supplier Thermo Scientific Chemicals Supplier No. J6104314

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A dye used to visualize beta-amyloid plaques, like those associated with Alzheimer's Disease | CAS: 2390-54-7 | C17H19ClN2S | 318.86 g/mol

A dye used to visualize beta-amyloid plaques, like those associated with Alzheimer's Disease

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2390-54-7
Molecular Formula C17H19ClN2S
Molecular Weight (g/mol) 318.86
MDL Number MFCD00011944
InChI Key JADVWWSKYZXRGX-UHFFFAOYSA-M
Synonym Basic Yellow 1; C.I. 49005
PubChem CID 16953
ChEBI CHEBI:76023
IUPAC Name 4-(3,6-dimethyl-1,3-benzothiazol-3-ium-2-yl)-N,N-dimethylaniline;chloride
SMILES [Cl-].CN(C)C1=CC=C(C=C1)C1=[N+](C)C2=CC=C(C)C=C2S1

Specifications

Beilstein 3922452
Color Yellow
Quantity 25 g
Physical Form Powder
Formula Weight 318.86
Percent Purity 75%
Grade Technical
Chemical Name or Material Thioflavine T
EINECSNumber 219-228-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only