missing translation for 'onlineSavingsMsg'
Learn More

Thiocyanate Standard, 1000 ppm SCN-, Ricca Chemical

$116.47 - $264.63

Chemical Identifiers

CAS 333-20-0
Molecular Formula CKNS
Molecular Weight (g/mol) 97.176
InChI Key ZNNZYHKDIALBAK-UHFFFAOYSA-M
PubChem CID 516872
ChEBI CHEBI:30951
IUPAC Name potassium;thiocyanate
SMILES C(#N)[S-].[K+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
8395-4
View Documents
Ricca Chemical Company
83954
120 mL Natural Poly Bottle
Each for $116.47
Only null left
 
8395-16
View Documents
Ricca Chemical Company
839516
500 mL Natural Poly Bottle
Each for $264.63
Only null left
 
Description

Description

Specifications

Chemical Identifiers

333-20-0
97.176
516872
potassium;thiocyanate
CKNS
ZNNZYHKDIALBAK-UHFFFAOYSA-M
CHEBI:30951
C(#N)[S-].[K+]

Specifications

Natural Poly Bottle
98.0,0.16
1 mL = 1 mg SCN
CKNS
100°C
C(#N)[S-].[K+]
97.176
516872
Thiocyanate Standard
333-20-0,7732-18-5
100.0,0.17
Liquid
Colorless
ZNNZYHKDIALBAK-UHFFFAOYSA-M
potassium;thiocyanate
120 mL
CHEBI:30951
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'