missing translation for 'onlineSavingsMsg'
Learn More

Thiocyanate Standard, 1000 ppm SCN-, Ricca Chemical

Catalog No. 83954
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
83954 120 mL Natural Poly Bottle
839516 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 83954 Supplier Ricca Chemical Company Supplier No. 83954

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 333-20-0 , 7732-18-5
Molecular Formula CKNS
Molecular Weight (g/mol) 97.176
InChI Key ZNNZYHKDIALBAK-UHFFFAOYSA-M
PubChem CID 516872
ChEBI CHEBI:30951
IUPAC Name potassium;thiocyanate
SMILES C(#N)[S-].[K+]

Specifications

Packaging Natural Poly Bottle
CAS Min % 98.0, 0.16
CAS Max % 100.0, 0.17
Equivalence 1 mL = 1 mg SCN
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Quantity 120 mL
Grade Not Available
Chemical Name or Material Thiocyanate Standard