missing translation for 'onlineSavingsMsg'
Learn More

Thioacetic acid, 98%

Catalog No. AC138785000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC138785000 500 g Glass bottle
AC138781000 100 g Glass bottle
AC138780025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC138785000 Supplier Thermo Scientific Chemicals Supplier No. 138785000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 507-09-5 | C2H4OS | 76.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 507-09-5
Molecular Formula C2H4OS
Molecular Weight (g/mol) 76.11
MDL Number MFCD00004853
InChI Key DUYAAUVXQSMXQP-UHFFFAOYSA-N
Synonym thioacetic acid, ethanethioic acid, thiacetic acid, thioacetic s-acid, acetyl mercaptan, ethanethiolic acid, thiolacetic acid, thionoacetic acid, methanecarbothiolic acid, ch3cosh
PubChem CID 10484
ChEBI CHEBI:16555
IUPAC Name ethanethioic S-acid
SMILES CC(S)=O

Specifications

Melting Point -17.0°C
Color Colorless to Yellow
Density 1.0700g/mL
Boiling Point 87.0°C to 93.0°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Refractive Index 1.4630 to 1.4650
Linear Formula CH3COSH
Quantity 500 g
Beilstein 02, 230
Fieser 01,1154
Specific Gravity 1.07
Merck Index 15, 9473
Solubility Information Solubility in water: 27g/L water (15°C) (hydrolyse). Other solubilities: soluble in alcohol and ether
Formula Weight 76.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Thioacetic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Toxic if swallowed.
Causes serious eye damage.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER
EINECSNumber 208-063-8
RTECSNumber AJ5600000
TSCA TSCA

RUO – Research Use Only