missing translation for 'onlineSavingsMsg'
Learn More

Thioacetamide, 0.27 Molar Unstandardized, Reagents

Catalog No. 01333460
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01333460 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01333460 Supplier Labchem Inc Supplier No. CT009500500A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thioacetamide, 0.27 Molar Unstandardized, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Thioacetamide, 0.27 Molar Unstandardized
CAS 7732-18-5, 62-55-5
Color Colorless to Slight Yellow
Boiling Point 100°C
Packaging Poly Bottle
Quantity 500 mL
Specific Gravity 1.02
Synonym Thiocarbamoyl Hydrazide, Aminothiourea, Hydrazinecarbothioamide
Grade Reagent
Physical Form Liquid