missing translation for 'onlineSavingsMsg'
Learn More

Thioacetamide, 0.25% (w/v), Ricca Chemical

Catalog No. p-3252562
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
83934 120 mL Natural Poly Bottle
839316 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 83934 Supplier Ricca Chemical Company Supplier No. 83934

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 62-55-5 , 7732-18-5
Molecular Formula C2H5NS
Molecular Weight (g/mol) 75.13
MDL Number MFCD00008070
InChI Key YUKQRDCYNOVPGJ-UHFFFAOYSA-N
Synonym thioacetamide, acetothioamide, thiacetamide, acetamide, thio, thioacetimidic acid, acetimidic acid, thio, usaf cb-21, usaf ek-1719, rcra waste number u218, acetic acid, thiono-, amide
PubChem CID 2723949
ChEBI CHEBI:32497
IUPAC Name ethanethioamide
SMILES CC(N)=S

Specifications

CAS Min % 98.0, 0.24
CAS Max % 100.0, 0.25
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Quantity 120 mL
Concentration or Composition Notes 0.25% (w/v)
Chemical Name or Material Thioacetamide