Polyester films just 25 mum thick, clearer and marginally pierceable with single-channel pipettes for sample recovery. Dimensions are 54.0 mm x 141.0 mm. Semi-pierceable, and leaves minimal residue.
Numéro de catalogue.
502388665
Fournisseur
Alkali ScientificCode fournisseur
SF32MLS
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Polyester films just 25 mum thick, clearer and marginally pierceable with single-channel pipettes for sample recovery. Dimensions are 54.0 mm x 141.0 mm. Semi-pierceable, and leaves minimal residue.