missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thimerosal Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 54-64-8 |
| Flash Point | 250°C - closed cup |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Linear Formula | 2-(C2H5 HgS)C6H4 CO2 Na |
| MDL Number | MFCD00013062 |
| Melting Point | 234°C to 237°C |
| Synonym | Thimerosal; 2-(Ethylmercuriomercapto)benzoic acid sodium salt; Ethylmercurithiosalicylic acid sodium salt; Mercury-([o-carboxyphenyl]thio)ethyl sodium salt; Sodium ethylmercurithiosalicylate |
| Type | Certified Reference Material |
| Quantity | 1 g |
| Formula Weight | 404.81 |
| Show More |