missing translation for 'onlineSavingsMsg'
Learn More

Thifensulfuron-methyl 97.0+%, TCI America™

Catalog No. T31665G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31665G 5 g
T31661G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T31665G Supplier TCI America Supplier No. T31665G
Only null left

Chemical Identifiers

CAS 79277-27-3
Molecular Formula C12H13N5O6S2
Molecular Weight (g/mol) 387.385
MDL Number MFCD00468118
InChI Key AHTPATJNIAFOLR-UHFFFAOYSA-N
Synonym thifensulfuron-methyl, harmony, thifensulfuron methyl, thiameturon-methyl, pinnacle, refine, thifensulfuronmethyl, caswell no. 573s, unii-5vfh25es6f, epa pesticide chemical code 128845
PubChem CID 73674
ChEBI CHEBI:83453
IUPAC Name methyl 3-[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)carbamoylsulfamoyl]thiophene-2-carboxylate
SMILES CC1=NC(=NC(=N1)OC)NC(=O)NS(=O)(=O)C2=C(SC=C2)C(=O)OC

Specifications

Melting Point 188°C
Color White-Yellow
Quantity 5 g
UN Number 3077
Formula Weight 387.39
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Thifensulfuron-methyl
RTECSNumber XM8463000
TSCA No