missing translation for 'onlineSavingsMsg'
Learn More

Thiazolidine-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. T13245G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13245G 5 g
T13241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T13245G Supplier TCI America Supplier No. T13245G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16310-13-7
Molecular Formula C4H7NO2S
Molecular Weight (g/mol) 133.165
MDL Number MFCD00068005
InChI Key ULSZVNJBVJWEJE-UHFFFAOYSA-N
Synonym thiazolidine-2-carboxylic acid, 1,3-thiazolane-2-carboxylic acid, 2-thiazolidinecarboxylic acid, beta-thiaproline, thiazolidine-2-carboxylicacid, acmc-20aiwa, thiazolidine carboxylic acid, 2-triazolidinecarboxylic acid, ulszvnjbvjweje-uhfffaoysa, tetrahydrothiazole-2-carboxylic acid
PubChem CID 42486
IUPAC Name 1,3-thiazolidine-2-carboxylic acid
SMILES C1CSC(N1)C(=O)O

Specifications

Melting Point 182°C
Color White
Quantity 5 g
Formula Weight 133.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Thiazolidine-2-carboxylic Acid